Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RICTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285439.1 | NP_001272368.1 | 35 | 4566 | ||
NM_152756.4 | NP_689969.2 | 34 | 4494 | |||
XM_011514006.2 | XP_011512308.1 | 34 | 4383 | |||
XM_017009311.1 | XP_016864800.1 | 34 | 4499 | |||
XM_017009312.1 | XP_016864801.1 | 33 | 4427 | |||
XM_017009313.1 | XP_016864802.1 | 34 | 4520 | |||
XM_017009314.1 | XP_016864803.1 | 34 | 4489 | |||
XM_017009315.1 | XP_016864804.1 | 33 | 4417 | |||
GenBank mRNA | AB082530.1 | - | 19 | 3165 | ||
AL834497.1 | - | 7 | 1744 | |||
AY515854.1 | - | 34 | 4490 | |||
BC051729.1 | - | 14 | 2492 | |||
BC137163.1 | - | 34 | 4487 | 9021160 | ||
BC137164.1 | - | 34 | 4487 | 9021161 | ||
BC144509.1 | - | 35 | 4559 | 9053036 |