Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014034.2 | NP_054753.1 | 4 | 832 | ||
GenBank mRNA | AB028628.1 | - | 4 | 658 | ||
AF151856.1 | - | 6 | 1109 | |||
AF161495.1 | - | 6 | 1109 | |||
AF279306.1 | - | 4 | 644 | |||
AK025738.1 | - | 4 | 689 | |||
AL050261.1 | - | 4 | 687 | |||
BC010878.1 | - | 4 | 671 | 3546858 | ||
CR457347.1 | - | 4 | 495 |