Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 3 | 1245 | ||
NM_018136.4 | NP_060606.3 | 3 | 1245 | |||
GenBank mRNA | AF509326.1 | - | 3 | 988 | ||
AK226178.1 | - | 3 | 1178 | |||
AY099892.1 | - | 3 | 988 | |||
AY101201.1 | - | 2 | 655 | |||
AY367065.1 | - | 3 | 988 | |||
AY971956.1 | - | 3 | 988 |