Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015621.2 | NP_056436.2 | 8 | 756 | ||
GenBank mRNA | AK023767.1 | - | 7 | 534 | ||
AK094658.1 | - | 1 | 792 | |||
AK225826.1 | - | 8 | 726 | |||
AK292841.1 | - | 8 | 756 | |||
AL713742.1 | - | 7 | 680 | |||
BC016647.1 | - | 7 | 569 | |||
BM927471.1 | - | 3 | 273 |