Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015621.2 | NP_056436.2 | 3 | 291 | ||
GenBank mRNA | AK023767.1 | - | 2 | 69 | ||
AK225826.1 | - | 3 | 261 | |||
AK292841.1 | - | 3 | 291 | |||
AK298063.1 | - | 3 | 290 | |||
AL713742.1 | - | 2 | 215 | |||
BC016647.1 | - | 2 | 129 |