Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015658.3 | NP_056473.2 | 2 | 152 | ||
GenBank mRNA | AK225239.1 | - | 2 | 107 | ||
AK315080.1 | - | 2 | 121 | |||
AL050019.1 | - | 2 | 108 | |||
BC003555.1 | - | 2 | 125 | 3546209 | ||
DA002961.1 | - | 2 | 152 |