Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014506.2 | NP_055321.1 | 5 | 879 | ||
NR_133948.1 | 5 | 1020 | ||||
NR_134027.1 | 3 | 437 | ||||
GenBank mRNA | AF007872.1 | - | 4 | 600 | ||
AF317129.1 | - | 5 | 869 | |||
BC015578.2 | - | 5 | 824 | 4589479 | ||
CN262366.1 | - | 4 | 669 | |||
HY036797.1 | - | 3 | 397 |