Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGSM3 SIRNA Assays ›
Gene Symbol:
SGSM3
Gene Name:
small G protein signaling modulator 3
Gene Aliases:
MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP
Chromosome Location:
Chr.22: 40370530 - 40410289 on Build GRCh38
UniGene ID:
Hs.474914
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s226124
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGSM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301849.1
NP_001288778.1
9
1068
NM_015705.5
NP_056520.2
10
1213
XM_005261572.2
XP_005261629.1
10
1240
XM_005261573.3
XP_005261630.1
10
1245
XM_005261574.4
XP_005261631.1
10
1245
XM_005261575.1
XP_005261632.1
9
1108
XM_011530158.1
XP_011528460.1
9
1064
XM_011530159.1
XP_011528461.1
10
1128
XM_011530160.1
XP_011528462.1
9
1045
XM_011530161.1
XP_011528463.1
10
1237
XM_011530162.2
XP_011528464.1
10
1123
XM_017028775.1
XP_016884264.1
9
1032
XM_017028776.1
XP_016884265.1
9
1101
XM_017028777.1
XP_016884266.1
9
1101
XM_017028778.1
XP_016884267.1
8
964
XM_017028779.1
XP_016884268.1
9
1154
XM_017028780.1
XP_016884269.1
9
1143
GenBank mRNA
AB196956.1
-
8
982
AB275763.1
-
9
1086
AB449880.1
-
8
975
AK000006.1
-
6
953
AK295469.1
-
10
1159
AK297599.1
-
9
1061
AK301042.1
-
9
981
AK309400.1
-
9
1383
AY308849.1
-
10
1148
BC008078.2
-
10
1139
3027777
BC127850.1
-
3
289
CR456441.1
-
9
1011
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
cell cycle arrest
regulation of vesicle fusion
regulation of Rab protein signal transduction
Rap protein signal transduction
plasma membrane to endosome transport
activation of GTPase activity
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline