Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085347.2 | NP_001078816.1 | 5 | 1303 | ||
NM_001134430.2 | NP_001127902.1 | 4 | 1175 | |||
NM_001134431.2 | NP_001127903.1 | 2 | 2015 | |||
NM_001252018.1 | NP_001238947.1 | 4 | 1037 | |||
NM_001252021.1 | NP_001238950.1 | 4 | 1034 | |||
NM_001252023.1 | NP_001238952.1 | 3 | 909 | |||
NM_130459.3 | NP_569726.2 | 3 | 2281 | |||
XM_011518554.1 | XP_011516856.1 | 5 | 1338 | |||
GenBank mRNA | AK027677.1 | - | 3 | 2216 | ||
AK055034.1 | - | 2 | 1961 | |||
AK075520.1 | - | 4 | 1118 | |||
AK092388.1 | - | 2 | 3247 | |||
AY358180.1 | - | 5 | 1270 | |||
BC022985.1 | - | 1 | 1175 | |||
BC062340.1 | - | 4 | 1002 | |||
BC080527.1 | - | 3 | 835 |