Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO80E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304562.1 | NP_001291491.1 | 3 | 289 | ||
NM_001304563.1 | NP_001291492.1 | 3 | 289 | |||
NM_173618.2 | NP_775889.1 | 3 | 289 | |||
NR_130786.1 | 3 | 289 | ||||
NR_130787.1 | 3 | 289 | ||||
NR_134855.1 | 3 | 289 | ||||
XM_011545809.2 | XP_011544111.1 | 3 | 273 | |||
XM_011545811.2 | XP_011544113.1 | 3 | 273 | |||
XM_011545812.2 | XP_011544114.1 | 3 | 273 | |||
XM_017023169.1 | XP_016878658.1 | 3 | 273 | |||
GenBank mRNA | AK074137.1 | - | 3 | 260 | ||
AK075133.1 | - | 3 | 288 | |||
AK130409.1 | - | 3 | 280 | |||
AK309498.1 | - | 3 | 253 | |||
AL525020.3 | - | 3 | 289 | |||
AY189289.1 | - | 3 | 224 | |||
BC035693.1 | - | 3 | 247 | |||
BC047712.2 | - | 3 | 250 | 6155713 | ||
BI093312.1 | - | 3 | 253 | |||
BM924992.1 | - | 250 | ||||
BU528528.1 | - | 3 | 250 | |||
BX356644.2 | - | 3 | 253 |