Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144939.1 | NP_001138411.1 | 2 | 368 | ||
NM_182566.2 | NP_872372.1 | 2 | 368 | |||
GenBank mRNA | AF521892.1 | - | 2 | 373 | ||
AY359099.1 | - | 2 | 298 | |||
BC104194.2 | - | 2 | 163 | 40031204 | ||
BC104195.1 | - | 2 | 336 | 40031205 |