Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014957.2 | NP_055772.2 | 8 | 1130 | ||
XM_005250838.4 | XP_005250895.2 | 8 | 1021 | |||
XM_005250839.2 | XP_005250896.2 | 8 | 1021 | |||
XM_005250840.4 | XP_005250897.2 | 8 | 1021 | |||
XM_005250841.3 | XP_005250898.2 | 8 | 1021 | |||
XM_005250842.4 | XP_005250899.1 | 6 | 882 | |||
XM_011516933.1 | XP_011515235.1 | 8 | 1021 | |||
XM_011516934.2 | XP_011515236.1 | 8 | 1021 | |||
XM_011516937.1 | XP_011515239.1 | 8 | 1021 | |||
XM_011516941.2 | XP_011515243.1 | 8 | 1021 | |||
XM_017013241.1 | XP_016868730.1 | 8 | 1023 | |||
XM_017013242.1 | XP_016868731.1 | 9 | 1384 | |||
GenBank mRNA | AB020677.2 | - | 3 | 317 | ||
BC133023.1 | - | 7 | 873 | 40147045 | ||
BC144226.1 | - | 7 | 873 | 9052746 |