Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321864.1 | NP_001308793.1 | 3 | 293 | ||
NM_001321865.1 | NP_001308794.1 | 3 | 419 | |||
NM_182575.2 | NP_872381.2 | 4 | 894 | |||
NR_135832.1 | 3 | 352 | ||||
XM_005258793.4 | XP_005258850.1 | 4 | 627 | |||
XM_011526811.2 | XP_011525113.1 | 3 | 1398 | |||
XM_011526812.2 | XP_011525114.1 | 4 | 974 | |||
XM_011526813.2 | XP_011525115.1 | 4 | 823 | |||
XM_011526815.2 | XP_011525117.1 | 3 | 1537 | |||
XM_017026646.1 | XP_016882135.1 | 3 | 1398 | |||
GenBank mRNA | AB195682.1 | - | 4 | 752 | ||
AK301750.1 | - | 3 | 1537 | |||
AY552609.1 | - | 3 | 346 | |||
AY552610.1 | - | 3 | 279 | |||
BC034769.1 | - | 4 | 894 | 5167853 |