Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320185.1 | NP_001307114.1 | 10, 11 | 1203 | ||
NM_181781.3 | NP_861446.2 | 8, 9 | 1006 | |||
NM_182976.3 | NP_892021.1 | 10, 11 | 1470 | |||
XM_005270779.4 | XP_005270836.1 | 9, 10 | 1044 | |||
XM_011541290.2 | XP_011539592.1 | 10, 11 | 1176 | |||
GenBank mRNA | AK297073.1 | - | 10, 11 | 1395 | ||
AK303707.1 | - | 8, 9 | 933 | |||
BC014899.1 | - | 10, 11 | 1161 | |||
BC070341.1 | - | 10, 11 | 1414 | |||
BC090866.1 | - | 10, 11 | 1464 | 6503526 | ||
BU521884.1 | - | 2, 3 | 214 |