Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025014.1 | NP_079290.1 | 13, 14 | 2275 | ||
NM_173728.3 | NP_776089.2 | 13, 14 | 2339 | |||
XM_011523734.1 | XP_011522036.1 | 13, 14 | 2384 | |||
XM_011523735.1 | XP_011522037.1 | 13, 14 | 2320 | |||
XM_011523736.1 | XP_011522038.1 | 13, 14 | 2335 | |||
XM_011523737.1 | XP_011522039.1 | 10, 11 | 1376 | |||
GenBank mRNA | AB020722.1 | - | 13, 14 | 2255 | ||
AK291626.1 | - | 13, 14 | 2299 | |||
AK300327.1 | - | 13, 14 | 1760 | |||
BC036749.1 | - | 13, 14 | 2294 | 5204409 | ||
BQ877184.1 | - | 13 | ||||
KU178557.1 | - | 12, 13 | 2184 |