Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR663AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040095.1 | 5 | 424 | |||
GenBank mRNA | BC036201.1 | - | 3 | 927 | ||
BC036544.1 | - | 5 | 424 |