Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046322.1 | 3 | 1514 | |||
GenBank mRNA | AK057419.1 | - | 3 | 986 | ||
BC101730.1 | - | 1 | 229 | 8069236 | ||
BC101732.1 | - | 1 | 229 | 8069238 |