Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARD8 SIRNA Assays ›
Gene Symbol:
CARD8
Gene Name:
caspase recruitment domain family member 8
Gene Aliases:
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location:
Chr.19: 48203148 - 48256269 on Build GRCh38
UniGene ID:
Hs.446146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22623
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184900.1
NP_001171829.1
5
492
NM_001184901.1
NP_001171830.1
5
439
NM_001184902.1
NP_001171831.1
8
855
NM_001184903.1
NP_001171832.1
6
589
NM_014959.3
NP_055774.2
6
550
NR_033678.1
4
412
NR_033679.1
4
451
NR_033680.1
4
412
XM_006723091.3
XP_006723154.1
7
703
XM_006723092.3
XP_006723155.1
9
1241
XM_006723093.3
XP_006723156.1
7
1024
XM_006723094.3
XP_006723157.1
7
686
XM_006723095.3
XP_006723158.1
7
670
XM_006723096.3
XP_006723159.1
8
1092
XM_006723097.3
XP_006723160.1
6
875
XM_006723098.3
XP_006723161.1
6
662
XM_006723099.3
XP_006723162.1
5
548
XM_006723101.2
XP_006723164.1
6
565
XM_006723102.3
XP_006723165.1
5
505
XM_006723104.3
XP_006723167.1
6
565
XM_006723106.3
XP_006723169.1
7
1035
XM_006723107.3
XP_006723170.1
7
686
XM_006723109.3
XP_006723172.1
6
565
XM_011526641.2
XP_011524943.1
10
1548
XM_011526642.2
XP_011524944.1
9
1443
XM_011526643.2
XP_011524945.1
8
1130
XM_011526644.2
XP_011524946.1
6
715
XM_011526646.2
XP_011524948.1
7
1189
XM_011526647.2
XP_011524949.1
6
1078
XM_011526648.2
XP_011524950.1
7
1198
XM_011526650.2
XP_011524952.1
7
686
XM_011526653.2
XP_011524955.1
7
1228
XM_017026482.1
XP_016881971.1
7
1031
XM_017026483.1
XP_016881972.1
7
981
XM_017026484.1
XP_016881973.1
7
992
XM_017026485.1
XP_016881974.1
7
686
XM_017026486.1
XP_016881975.1
5
548
XM_017026487.1
XP_016881976.1
4
914
XM_017026488.1
XP_016881977.1
8
1130
XM_017026489.1
XP_016881978.1
7
715
XM_017026490.1
XP_016881979.1
7
686
XM_017026491.1
XP_016881980.1
7
686
XM_017026492.1
XP_016881981.1
7
997
XM_017026493.1
XP_016881982.1
6
889
XM_017026494.1
XP_016881983.1
7
1503
XM_017026495.1
XP_016881984.1
6
645
XM_017026496.1
XP_016881985.1
6
565
XM_017026497.1
XP_016881986.1
5
505
XM_017026498.1
XP_016881987.1
5
505
XM_017026499.1
XP_016881988.1
4
2278
GenBank mRNA
AB023172.1
-
4
447
AF322184.1
-
4
447
AF331519.1
-
2
134
AF405558.1
-
2
134
AF511652.1
-
4
447
AK297045.1
-
6
575
AK297069.1
-
5
429
AK302327.1
-
4
379
AY026322.1
-
2
134
BX641091.1
-
1
362
BX648568.1
-
6
2596
EU118120.1
-
5
492
EU118121.1
-
4
451
EU118122.1
-
2
292
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of interleukin-1 beta secretion
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
NACHT domain binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline