Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM176B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101311.1 | NP_001094781.1 | 6 | 821 | ||
NM_001101312.1 | NP_001094782.1 | 5 | 813 | |||
NM_001101314.1 | NP_001094784.1 | 4 | 702 | |||
NM_014020.3 | NP_054739.3 | 5 | 945 | |||
XM_006715933.3 | XP_006715996.2 | 5 | 589 | |||
GenBank mRNA | AF115384.1 | - | 5 | 735 | ||
AK000841.1 | - | 5 | 644 | |||
AK097304.1 | - | 6 | 1782 | |||
AK129525.1 | - | 4 | 538 | |||
AK315575.1 | - | 6 | 631 | |||
BC004358.1 | - | 5 | 860 | 2822223 | ||
BC014586.2 | - | 5 | 860 | 2822223 | ||
BC091471.1 | - | 5 | 644 | |||
BI759037.1 | - | 4 | 425 | |||
BI761752.1 | - | 5 | 815 | |||
BI910761.1 | - | 6 | 617 | |||
DQ144974.1 | - | 5 | 703 |