Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318817.1 | NP_001305746.1 | 2 | 242 | ||
NM_014161.4 | NP_054880.2 | 2 | 242 | |||
NR_134860.1 | 3 | 362 | ||||
NR_134861.1 | 2 | 319 | ||||
NR_134862.1 | 2 | 242 | ||||
NR_134863.1 | 2 | 242 | ||||
NR_134864.1 | 2 | 125 | ||||
NR_134865.1 | 2 | 242 | ||||
NR_134866.1 | 2 | 242 | ||||
GenBank mRNA | AF161556.1 | - | 2 | 220 | ||
AK311391.1 | - | 3 | 239 | |||
AL136633.1 | - | 2 | 192 | |||
AV756740.1 | - | 2 | 177 | |||
BC001623.1 | - | 2 | 213 | 3357333 | ||
BF797811.1 | - | 2 | 125 | |||
BG548185.1 | - | 2 | 221 | |||
BM924328.1 | - | 2 | 335 | |||
BU601065.1 | - | 2 | 183 | |||
BU932963.1 | - | 2 | 231 | |||
CA777760.1 | - | 2 | 124 | |||
CD675657.1 | - | 2 | 141 | |||
CR533491.1 | - | 2 | 100 |