Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130015.1 | NP_001123487.1 | 6 | 1617 | ||
NM_001304459.1 | NP_001291388.1 | 6 | 1880 | |||
NM_001304460.1 | NP_001291389.1 | 5 | 1491 | |||
NM_182608.3 | NP_872414.3 | 5 | 1791 | |||
GenBank mRNA | AF517120.1 | - | 1 | 374 | ||
AK125517.1 | - | 5 | 1840 | |||
BX537908.1 | - | 6 | 1392 | |||
BX640656.1 | - | 6 | 1392 | |||
BX647684.1 | - | 2 | 3091 |