Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 6 | 560 | |||
NR_027791.3 | 5 | 478 | ||||
NR_111005.2 | 6 | 605 | ||||
NR_136544.1 | 3 | 630 | ||||
NR_136545.1 | 3 | 630 | ||||
NR_136547.1 | 2 | 180 | ||||
NR_136550.1 | 3 | 369 | ||||
NR_136551.1 | 5 | 513 | ||||
NR_136552.1 | 5 | 513 | ||||
GenBank mRNA | AK095614.1 | - | 6 | 560 | ||
AL700268.1 | - | 5 | 389 |