Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PINLYP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193621.1 | NP_001180550.1 | 5 | 657 | ||
NM_001193622.1 | NP_001180551.1 | 3 | 317 | |||
NM_001321124.1 | NP_001308053.1 | 3 | 337 | |||
XM_011526970.2 | XP_011525272.1 | 4 | 2253 | |||
XM_011526971.2 | XP_011525273.1 | 3 | 679 | |||
GenBank mRNA | AK129550.1 | - | 3 | 305 | ||
BC071811.1 | - | 3 | 317 | |||
HY005068.1 | - | 3 | 337 |