Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007367.3 | NP_031393.2 | 5 | 1000 | ||
NM_016732.2 | NP_057951.1 | 6 | 1048 | |||
XM_005260334.4 | XP_005260391.1 | 5 | 963 | |||
XM_005260336.4 | XP_005260393.1 | 4 | 915 | |||
XM_011528694.2 | XP_011526996.1 | 6 | 606 | |||
XM_011528695.2 | XP_011526997.1 | 7 | 1158 | |||
XM_017027731.1 | XP_016883220.1 | 5 | 963 | |||
GenBank mRNA | AF148457.1 | - | 6 | 774 | ||
AK222915.1 | - | 6 | 734 | |||
AK226085.1 | - | 5 | 899 | |||
AK291079.1 | - | 6 | 625 | |||
AY635580.1 | - | 1 | 27 | |||
BC103753.1 | - | 6 | 795 | 6182358 | ||
BC105018.1 | - | 6 | 643 | 8144021 | ||
L38696.1 | - | 5 | 604 |