Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 703 | ||
NM_175868.2 | NP_787064.1 | 3 | 767 | |||
GenBank mRNA | AK289437.1 | - | 3 | 687 | ||
BC041599.1 | - | 3 | 676 | 5458512 | ||
BC067731.1 | - | 3 | 684 | 4828429 | ||
BE542284.1 | - | 3 | 661 | |||
BX640600.1 | - | 3 | 1064 | |||
D32076.1 | - | 1 | 561 | |||
EU161101.1 | - | 1 | 496 | |||
U10339.1 | - | 1 | 496 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005362.3 | NP_005353.1 | 3 | 705 | ||
XM_005274676.3 | XP_005274733.1 | 3 | 779 | |||
XM_006724818.2 | XP_006724881.1 | 3 | 861 | |||
XM_011531160.2 | XP_011529462.1 | 3 | 685 | |||
XM_011531161.2 | XP_011529463.1 | 3 | 1110 | |||
GenBank mRNA | AK223519.1 | - | 1 | 532 | ||
AK292384.1 | - | 3 | 705 | |||
BC000340.2 | - | 3 | 670 | 2822978 | ||
BC005963.1 | - | 3 | 705 | 4064879 | ||
BC011744.2 | - | 2 | 631 | 3345801 | ||
BC016803.1 | - | 3 | 667 | 4073332 | ||
BC017389.1 | - | 3 | 662 | 4047542 | ||
CR541767.1 | - | 1 | 496 | |||
CR541768.1 | - | 1 | 496 | |||
CR541774.1 | - | 1 | 496 | |||
CR542020.1 | - | 1 | 496 |