Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPV17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002437.4 | NP_002428.1 | 8 | 697 | ||
XM_005264326.3 | XP_005264383.1 | 8 | 709 | |||
XM_006712021.3 | XP_006712084.1 | 9 | 849 | |||
XM_017004150.1 | XP_016859639.1 | 9 | 3926 | |||
XM_017004151.1 | XP_016859640.1 | 8 | 745 | |||
XM_017004152.1 | XP_016859641.1 | 8 | 768 | |||
GenBank mRNA | AK093305.1 | - | 6 | 1703 | ||
BC001115.2 | - | 7 | 688 | 2967261 | ||
BC016289.2 | - | 7 | 631 | |||
BG714998.1 | - | 6 | 508 | |||
X76538.1 | - | 8 | 670 |