Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038857.1 | 2 | 123 | |||
GenBank mRNA | BC039114.1 | - | 2 | 123 | ||
BC044764.1 | - | 2 | 123 | |||
BC047127.1 | - | 2 | 123 | |||
DB058059.1 | - | 2 | 110 | |||
DB089460.1 | - | 2 | 71 |