Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005952.3 | NP_005943.1 | 3 | 228 | ||
GenBank mRNA | BC018190.1 | - | 3 | 213 | 3859643 | |
BC032131.2 | - | 3 | 373 | 4053295 | ||
BC032338.1 | - | 3 | 215 | 5190391 | ||
CD101947.1 | - | 3 | 232 | |||
X76717.1 | - | 3 | 209 |