Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015675.3 | NP_056490.2 | 4 | 1136 | ||
XM_017026822.1 | XP_016882311.1 | 3 | 1370 | |||
GenBank mRNA | AF087853.1 | - | 4 | 1124 | ||
AF090950.1 | - | 5 | 804 | |||
AK129595.1 | - | 1 | 1159 | |||
AL050044.1 | - | 1 | 289 |