Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320227.1 | NP_001307156.1 | 5 | 480 | ||
NM_015938.4 | NP_057022.2 | 5 | 480 | |||
XM_005247511.1 | XP_005247568.1 | 5 | 430 | |||
XM_005247512.1 | XP_005247569.1 | 5 | 761 | |||
GenBank mRNA | AF132941.1 | - | 5 | 451 | ||
AK021775.1 | - | 2 | 91 | |||
AK094749.1 | - | 4 | 783 | |||
AK225664.1 | - | 5 | 462 | |||
AK308865.1 | - | 5, 6 | 459 | |||
BC013317.1 | - | 5 | 463 | 4291797 |