Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321329.1 | NP_001308258.1 | 3 | 601 | ||
NM_001321330.1 | NP_001308259.1 | 2 | 226 | |||
NM_001321331.1 | NP_001308260.1 | 3 | 234 | |||
NM_001321332.1 | NP_001308261.1 | 3 | 227 | |||
NM_001321333.1 | NP_001308262.1 | 3 | 234 | |||
NM_016133.3 | NP_057217.2 | 3 | 608 | |||
GenBank mRNA | AF125392.1 | - | 2 | 522 | ||
AF527632.1 | - | 3 | 557 | |||
AK021692.1 | - | 3 | 139 | |||
AK291433.1 | - | 3 | 556 | |||
AK298599.1 | - | 2 | 190 | |||
BC022475.1 | - | 3 | 552 | 4794938 | ||
BE888848.1 | - | 3 | 230 | |||
BX647805.1 | - | 3 | 149 | |||
DC364733.1 | - | 2 | 191 |