Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005018.2 | NP_005009.2 | 5 | 1114 | ||
XM_017004293.1 | XP_016859782.1 | 5 | 1113 | |||
GenBank mRNA | U64863.1 | - | 5 | 1114 |