Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016337.2 | NP_057421.1 | 4, 5 | 503 | ||
XM_005267749.3 | XP_005267806.1 | 4, 5 | 1122 | |||
XM_011536828.2 | XP_011535130.1 | 4, 5 | 1136 | |||
XM_017021363.1 | XP_016876852.1 | 4, 5 | 1122 | |||
XM_017021364.1 | XP_016876853.1 | 4, 5 | 1009 | |||
GenBank mRNA | AF052504.1 | - | 4, 5 | 477 | ||
AF087843.1 | - | 4, 5 | 482 | |||
AF112209.1 | - | 4, 5 | 596 | |||
AF131766.1 | - | 2, 3 | 250 | |||
AK289720.1 | - | 5, 6 | 513 | |||
AK295919.1 | - | 4, 5 | 711 | |||
AK301379.1 | - | 4, 5 | 495 | |||
BC023997.2 | - | 4, 5 | 652 | 2822488 | ||
BC032358.1 | - | 4, 5 | 474 | 5222624 | ||
BQ083568.1 | - | 4, 5 | 503 |