Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286229.1 | NP_001273158.1 | 12 | 1748 | ||
NM_001286230.1 | NP_001273159.1 | 13 | 1852 | |||
NM_016448.3 | NP_057532.3 | 14 | 1978 | |||
XM_011509614.1 | XP_011507916.1 | 14 | 1682 | |||
GenBank mRNA | AF195765.1 | - | 14 | 1742 | ||
AF345896.1 | - | 14 | 1763 | |||
AK000742.1 | - | 14 | 1796 | |||
AK001206.1 | - | 1 | 238 | |||
AK001261.1 | - | 5 | 785 | |||
AK027651.1 | - | 12 | 1455 | |||
AK292343.1 | - | 14 | 1850 | |||
AK299649.1 | - | 4 | 775 | |||
AK303346.1 | - | 13 | 1655 | |||
BC033297.1 | - | 14 | 1719 | 4342989 | ||
BC033540.1 | - | 14 | 1760 | 4826434 | ||
DQ641253.1 | - | 14 | 1640 |