Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286705.1 | NP_001273634.1 | 2 | 199 | ||
NM_001286706.1 | NP_001273635.1 | 2 | 199 | |||
NM_016617.3 | NP_057701.1 | 2 | 199 | |||
NR_104584.1 | 2 | 199 | ||||
NR_104585.1 | 2 | 199 | ||||
GenBank mRNA | AB154404.1 | - | 1 | 50 | ||
AF208844.1 | - | 2 | 89 | |||
AK095490.1 | - | 2 | 119 | |||
AK307111.1 | - | 2 | 119 | |||
AK309914.1 | - | 2 | 85 | |||
AL515797.3 | - | 1 | 51 | |||
BC005193.1 | - | 2 | 116 | 3829206 | ||
BG540094.1 | - | 2 | 87 | |||
BG717690.1 | - | 2 | 121 | |||
BI458294.1 | - | 2 | 170 | |||
BP277186.1 | - | 2 | 199 | |||
BU589105.1 | - | 2 | 115 | |||
CR457189.1 | - | 1 | 50 | |||
DA448931.1 | - | 2 | 112 | |||
DA483023.1 | - | 2 | 119 | |||
Z70222.1 | - | 2 | 90 |