Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270879.1 | NP_001257808.1 | 4 | 497 | ||
NM_016098.3 | NP_057182.1 | 3 | 281 | |||
NR_073093.1 | 4 | 510 | ||||
XM_011535894.1 | XP_011534196.1 | 3 | 1280 | |||
XM_011535895.1 | XP_011534197.1 | 2 | 1064 | |||
GenBank mRNA | AF125101.1 | - | 3 | 266 | ||
AF151887.1 | - | 3 | 263 | |||
AK312201.1 | - | 3 | 205 | |||
AL556427.3 | - | 3 | 200 | |||
AL561589.3 | - | 4 | 385 | |||
BC000810.1 | - | 3 | 191 | 3452973 | ||
BF665465.1 | - | 4 | 369 | |||
BG828174.1 | - | 3 | 239 |