Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271880.1 | NP_001258809.1 | 4 | 519 | ||
NM_016438.3 | NP_057522.1 | 3 | 603 | |||
NR_073504.1 | 2 | 468 | ||||
XM_006721946.2 | XP_006722009.1 | 4 | 497 | |||
XM_006721947.3 | XP_006722010.1 | 4 | 907 | |||
XM_006721948.3 | XP_006722011.1 | 4 | 601 | |||
XM_011524891.2 | XP_011523193.1 | 4 | 1001 | |||
XM_017024742.1 | XP_016880231.1 | 4 | 947 | |||
XM_017024743.1 | XP_016880232.1 | 4 | 1039 | |||
GenBank mRNA | AB095269.1 | - | 1 | 153 | ||
BC020667.1 | - | 3 | 594 | 4724613 | ||
BM876435.1 | - | 2 | 444 |