Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123383.1 | NP_001116855.1 | 4 | 1942 | ||
NM_001123384.1 | NP_001116856.1 | 4 | 1942 | |||
NM_001123385.1 | NP_001116857.1 | 4 | 1871 | |||
NM_017745.5 | NP_060215.4 | 4 | 1871 | |||
XM_005272616.1 | XP_005272673.1 | 4 | 2003 | |||
XM_005272618.3 | XP_005272675.1 | 4 | 1669 | |||
XM_005272619.3 | XP_005272676.1 | 4 | 2329 | |||
XM_005272620.3 | XP_005272677.1 | 4 | 2329 | |||
XM_006724536.3 | XP_006724599.1 | 4 | 3105 | |||
XM_011543929.2 | XP_011542231.1 | 4 | 1779 | |||
XM_011543930.1 | XP_011542232.1 | 4 | 1792 | |||
XM_011543931.2 | XP_011542233.1 | 4 | 1902 | |||
XM_017029615.1 | XP_016885104.1 | 4 | 3105 | |||
XM_017029616.1 | XP_016885105.1 | 4 | 2329 | |||
GenBank mRNA | AB046795.2 | - | 3 | 1597 | ||
AF317391.1 | - | 4 | 1830 | |||
AF317392.1 | - | 4 | 1830 | |||
AK056538.1 | - | 4 | 1696 | |||
AK074286.1 | - | 1 | 932 | |||
AY316592.1 | - | 4 | 1830 | |||
BC114220.1 | - | 4 | 1942 | 4384724 | ||
BC128456.1 | - | 3 | 1607 |