Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012228.3 | NP_036360.3 | 5 | 612 | ||
GenBank mRNA | AF122004.1 | - | 5 | 529 | ||
AF151889.1 | - | 5 | 560 | |||
BC018030.1 | - | 5 | 607 | 4799583 | ||
BC066305.1 | - | 5 | 526 | |||
BC117471.1 | - | 5 | 661 | 40126022 | ||
BC130380.1 | - | 5 | 661 | 40146411 |