Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012228.3 | NP_036360.3 | 4 | 461 | ||
GenBank mRNA | AF122004.1 | - | 4 | 378 | ||
BC018030.1 | - | 4 | 456 | 4799583 | ||
BC066305.1 | - | 4 | 375 | |||
BC117471.1 | - | 4 | 510 | 40126022 | ||
BC130380.1 | - | 4 | 510 | 40146411 |