Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRASLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017878.1 | NP_060348.1 | 2 | 106 | ||
XM_011545120.2 | XP_011543422.1 | 3 | 558 | |||
GenBank mRNA | AB453251.1 | - | 1 | 47 | ||
AK000563.1 | - | 2 | 106 |