Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032142.3 | NP_115518.3 | 30 | 5662 | ||
XM_005258107.3 | XP_005258164.1 | 30 | 5717 | |||
XM_005258109.3 | XP_005258166.1 | 29 | 5507 | |||
XM_005258110.1 | XP_005258167.1 | 17 | 3901 | |||
XM_006722326.3 | XP_006722389.1 | 30 | 5717 | |||
XM_006722327.3 | XP_006722390.1 | 29 | 5507 | |||
XM_006722330.3 | XP_006722393.1 | 24 | 4700 | |||
XM_011525673.2 | XP_011523975.1 | 29 | 5543 | |||
XM_011525675.2 | XP_011523977.1 | 27 | 5266 | |||
XM_017025803.1 | XP_016881292.1 | 29 | 5552 | |||
XM_017025804.1 | XP_016881293.1 | 24 | 4700 | |||
XM_017025805.1 | XP_016881294.1 | 19 | 4041 | |||
XM_017025806.1 | XP_016881295.1 | 27 | 5147 | |||
XM_017025807.1 | XP_016881296.1 | 23 | 4759 | |||
GenBank mRNA | AB046789.1 | - | 11 | 2841 | ||
AF318363.1 | - | 10 | 1226 | |||
AK074074.1 | - | 15 | 3166 | |||
AK094448.1 | - | 1 | 36 | |||
AL136818.1 | - | 15 | 3315 | |||
AL833494.2 | - | 21 | 4380 | |||
BC144480.1 | - | 20 | 4198 | 9053007 | ||
BC144481.1 | - | 21 | 4408 | 9053008 | ||
BX648232.1 | - | 24 | 6321 | |||
BX649151.1 | - | 17 | 3901 | |||
KJ567064.1 | - | 29 | 5582 |