Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018193.2 | NP_060663.2 | 23, 24 | 2554 | ||
XM_011521764.2 | XP_011520066.1 | 23, 24 | 2578 | |||
GenBank mRNA | AK001581.1 | - | 17, 18 | 1943 | ||
AK027564.1 | - | 12, 13 | 1441 | |||
AK055176.1 | - | 23, 24 | 2513 |