Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278211.1 | NP_001265140.1 | 5 | 1591 | ||
NM_001278212.1 | NP_001265141.1 | 5 | 1536 | |||
NM_001278213.1 | NP_001265142.1 | 4 | 1335 | |||
NM_001278214.1 | NP_001265143.1 | 4 | 1391 | |||
NM_018205.3 | NP_060675.1 | 4 | 1368 | |||
NM_018239.3 | NP_060709.2 | 4 | 1386 | |||
NM_207119.2 | NP_997002.1 | 5 | 1849 | |||
NR_103467.1 | 3 | 1241 | ||||
NR_103468.1 | 3 | 1223 | ||||
XM_017016374.1 | XP_016871863.1 | 5 | 1531 | |||
XM_017016375.1 | XP_016871864.1 | 5 | 1526 | |||
GenBank mRNA | AK001613.1 | - | 4 | 1345 | ||
AK001706.1 | - | 4 | 1361 | |||
AK094734.1 | - | 5 | 1532 | |||
AL122117.1 | - | 2 | 1073 | |||
AY358318.1 | - | 5 | 1593 | |||
BC024001.2 | - | 5 | 1527 | 4822655 |