Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304529.1 | NP_001291458.1 | 2 | 174 | ||
NM_001304530.1 | NP_001291459.1 | 2 | 174 | |||
NM_001304532.1 | NP_001291461.1 | 2 | 174 | |||
NM_018246.2 | NP_060716.2 | 2 | 174 | |||
NR_130761.1 | 2 | 174 | ||||
NR_130762.1 | 2 | 174 | ||||
GenBank mRNA | AK001715.1 | - | 2 | 174 | ||
AK027900.1 | - | 2 | 126 | |||
AK292875.1 | - | 2 | 161 | |||
AK294848.1 | - | 2 | 161 | |||
AK311437.1 | - | 2 | 137 | |||
AK316015.1 | - | 2 | 128 | |||
BC006239.2 | - | 2 | 101 | |||
BC032588.1 | - | 2 | 163 | 5502227 | ||
BC049197.1 | - | 2 | 128 | 5534448 | ||
BC054039.1 | - | 2 | 114 | |||
BC065295.1 | - | 2 | 115 | 6139413 | ||
BC110391.1 | - | 2 | 115 | |||
BG718771.1 | - | 2 | 166 | |||
BU602778.1 | - | 2 | 175 | |||
DA218015.1 | - | 2 | 174 |