Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308277.1 | NP_001295206.1 | 8 | 1894 | ||
NM_018381.3 | NP_060851.2 | 8 | 2002 | |||
XM_011528121.1 | XP_011526423.1 | 10 | 2172 | |||
XM_017026934.1 | XP_016882423.1 | 8 | 1940 | |||
GenBank mRNA | AK002148.1 | - | 8 | 1788 | ||
AK096142.1 | - | 8 | 2017 | |||
AK098062.1 | - | 1 | 1530 | |||
AL137694.1 | - | 7 | 1826 |