Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307943.1 | NP_001294872.1 | 11 | 1456 | ||
NM_018259.5 | NP_060729.2 | 11 | 1456 | |||
XM_005253016.1 | XP_005253073.1 | 11 | 1456 | |||
XM_006718263.1 | XP_006718326.1 | 9 | 1139 | |||
XM_011520218.2 | XP_011518520.1 | 11 | 1456 | |||
XM_017018010.1 | XP_016873499.1 | 11 | 1456 | |||
GenBank mRNA | AK001540.1 | - | 6 | 691 | ||
AK001752.1 | - | 11 | 1409 | |||
BC017238.2 | - | 11 | 1405 | 3940012 | ||
BC033000.1 | - | 11 | 1402 | 4830377 | ||
BX640972.1 | - | 6 | 1296 |