Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159643.1 | NP_001153115.1 | 8 | 1223 | ||
NM_001159644.1 | NP_001153116.1 | 1 | 355 | |||
NM_018349.3 | NP_060819.3 | 8 | 1223 | |||
XM_005254955.3 | XP_005255012.1 | 9 | 1704 | |||
XM_005254960.2 | XP_005255017.1 | 1 | 360 | |||
XM_006720603.2 | XP_006720666.1 | 10 | 1460 | |||
XM_011521770.1 | XP_011520072.1 | 8 | 1223 | |||
XM_011521771.2 | XP_011520073.1 | 10 | 1629 | |||
XM_011521772.2 | XP_011520074.1 | 9 | 1519 | |||
XM_011521773.2 | XP_011520075.1 | 8 | 1223 | |||
XM_011521774.2 | XP_011520076.1 | 8 | 1223 | |||
XM_011521775.2 | XP_011520077.1 | 2 | 975 | |||
XM_017022403.1 | XP_016877892.1 | 7 | 1187 | |||
XM_017022404.1 | XP_016877893.1 | 10 | 1629 | |||
XM_017022405.1 | XP_016877894.1 | 9 | 1634 | |||
GenBank mRNA | AK002037.1 | - | 1 | 63 | ||
AK090622.1 | - | 9 | 1312 | |||
AK300325.1 | - | 9 | 1350 | |||
AY656717.1 | - | 8 | 1158 | |||
BC025708.1 | - | 1 | 316 | |||
BC041387.1 | - | 10 | 1646 | 5276211 | ||
BC111024.1 | - | 1 | 188 | |||
BC131527.1 | - | 8 | 1158 | 40035019 | ||
DA862691.1 | - | 1 | 360 |