Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018467.3 | NP_060937.1 | 5, 6 | 462 | ||
XM_017026975.1 | XP_016882464.1 | 6, 7 | 1172 | |||
XM_017026976.1 | XP_016882465.1 | 4, 5 | 412 | |||
GenBank mRNA | AB097050.1 | - | 5, 6 | 433 | ||
AF220052.1 | - | 5, 6 | 433 | |||
AK074683.1 | - | 5 | 1321 | |||
BC006005.1 | - | 5, 6 | 418 | 4279653 | ||
BC008455.1 | - | 5, 6 | 410 | 4247241 | ||
BI758045.1 | - | 5, 6 | 449 |