Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 11 | 923 | ||
NM_018491.3 | NP_060961.3 | 10 | 825 | |||
XM_005251513.4 | XP_005251570.1 | 9 | 773 | |||
XM_005251514.1 | XP_005251571.1 | 9 | 738 | |||
XM_011517962.2 | XP_011516264.1 | 10 | 825 | |||
XM_011517963.1 | XP_011516265.1 | 7 | 363 | |||
GenBank mRNA | AF257330.2 | - | 10 | 783 | ||
AK290648.1 | - | 11 | 895 | |||
AK315855.1 | - | 10 | 749 | |||
AY343911.1 | - | 10 | 782 | |||
BC005996.1 | - | 10 | 825 | 4069589 | ||
BC009573.1 | - | 11 | 876 | 4133536 | ||
BC013432.1 | - | 10 | 796 | 4133722 | ||
BC132759.1 | - | 10 | 759 | 40146781 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 10 | 897 | ||
XM_005263602.2 | XP_005263659.1 | 9 | 810 | |||
XM_005263603.2 | XP_005263660.1 | 11 | 931 | |||
XM_011510667.1 | XP_011508969.1 | 10 | 897 | |||
XM_011510668.2 | XP_011508970.1 | 7 | 457 | |||
XM_011510669.2 | XP_011508971.1 | 11 | 941 | |||
XM_017003408.1 | XP_016858897.1 | 12 | 1061 | |||
GenBank mRNA | AF452722.1 | - | 10 | 753 | ||
AK124867.1 | - | 1 | 2462 | |||
AK225251.1 | - | 11 | 911 | |||
AK290072.1 | - | 10 | 897 | |||
AY343910.1 | - | 10 | 782 | |||
BC120992.1 | - | 10 | 768 | 40116411 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291821.1 | NP_001278750.1 | 9 | 746 | ||
NM_201453.3 | NP_958861.2 | 10 | 806 | |||
XM_005277639.2 | XP_005277696.1 | 9 | 798 | |||
XM_006717116.1 | XP_006717179.1 | 10 | 885 | |||
XM_011518706.2 | XP_011517008.1 | 5 | 336 | |||
XM_017014730.1 | XP_016870219.1 | 8 | 738 | |||
XM_017014731.1 | XP_016870220.1 | 10 | 3257 | |||
XM_017014732.1 | XP_016870221.1 | 4 | 249 | |||
XM_017014733.1 | XP_016870222.1 | 5 | 5556 | |||
GenBank mRNA | AK297912.1 | - | 9 | 708 | ||
AY344633.1 | - | 10 | 782 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286835.1 | NP_001273764.1 | 12 | 1970 | ||
XM_005272748.1 | XP_005272805.1 | 10 | 822 | |||
XM_005272750.1 | XP_005272807.1 | 9 | 762 | |||
XM_005272751.1 | XP_005272808.1 | 9 | 735 | |||
XM_006716914.1 | XP_006716977.1 | 10 | 822 | |||
XM_017014455.1 | XP_016869944.1 | 9 | 735 | |||
XM_017014456.1 | XP_016869945.1 | 11 | 753 | |||
XM_017014457.1 | XP_016869946.1 | 10 | 666 | |||
XM_017014459.1 | XP_016869948.1 | 12 | 1001 | |||
XM_017014461.1 | XP_016869950.1 | 11 | 914 | |||
GenBank mRNA | AK097639.1 | - | 10 | 1355 | ||
AK292585.1 | - | 11 | 890 | |||
BC067803.1 | - | 12 | 1908 | 5294979 | ||
BC141856.1 | - | 11 | 888 |
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085457.1 | NP_001078926.1 | 10 | 825 | ||
XM_017015019.1 | XP_016870508.1 | 9 | 825 | |||
XM_017015020.1 | XP_016870509.1 | 9 | 825 | |||
XM_017015022.1 | XP_016870511.1 | 8 | 738 | |||
XM_017015024.1 | XP_016870513.1 | 8 | 738 | |||
XM_017015026.1 | XP_016870515.1 | 9 | 825 | |||
XM_017015027.1 | XP_016870516.1 | 9 | 825 | |||
XM_017015028.1 | XP_016870517.1 | 10 | 693 | |||
XM_017015029.1 | XP_016870518.1 | 6 | 644 | |||
XM_017015031.1 | XP_016870520.1 | 5 | 336 | |||
XM_017015033.1 | XP_016870522.1 | 4 | 249 | |||
XM_017015034.1 | XP_016870523.1 | 4 | 199 | |||
GenBank mRNA | AF293368.1 | - | 3 | 130 | ||
AK293173.1 | - | 10 | 873 | |||
BC069006.1 | - | 11 | 966 |